Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dithiothreitol, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
100g
25g
5g
Packaging:
Poly Bottle
POLYSTORMOR

Chemical Identifiers

3483-12-3
C4H10O2S2
154.24
VHJLVAABSRFDPM-IMJSIDKUSA-N
DTT; Cleland's Reagent
446094
CHEBI:42170

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Laboratory,Research,or Manufacturing Use
Specific Gravity 1L = 1.00kg
Flash Point 109.00°C
Quantity 5g
CAS 3483-12-3
Packaging POLYSTORMOR
Molecular Formula C4H10O2S2
Synonym DTT; Cleland's Reagent
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
SMILES C(C(C(CS)O)O)S
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Molecular Weight (g/mol) 154.24
PubChem CID 446094
ChEBI CHEBI:42170
Formula Weight 154.25
Chemical Name or Material 1,4-Dithiothreitol
Grade Ultrapure Bioreagent
Show More Show Less