Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(4-Ethoxyphenyl)ethanol, 98%

Manufacturer:  Alfa Aesar™ H3005214

Quantity 25g

 View more versions of this product

Catalog No. AAH3005214


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

52067-36-4
166.22
1-(4-Ethoxyphenyl)ethanol
C10H14O2
MFCD08062382

Specifications

46° to 47°C
52067-36-4
166.22
1-(4-Ethoxyphenyl)ethanol
25g
C10H14O2
MFCD08062382
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit