Alfa Aesar™ 1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene, 99+%

Manufacturer: Alfa Aesar™ H2747609

Quantity 10g

 View more versions of this product

Catalog No. AAH2747609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 95480-29-8
    Molecular Formula C21H24O
    Formula Weight 292.42
    MDL Number MFCD01218020
    Synonym 1-(4-Ethoxyphenyl)-2-(4-n-pentylphenyl)acetylene
    Chemical Name or Material 1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene

    Specifications

    Formula Weight 292.42
    Melting Point 62°C
    Molecular Formula C21H24O
    MDL Number MFCD01218020
    CAS 95480-29-8
    Synonym 1-(4-Ethoxyphenyl)-2-(4-n-pentylphenyl)acetylene
    Boiling Point 178° to 180°C (0.5mmHg)
    Quantity 10g
    Chemical Name or Material 1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit