Alfa Aesar™ 1-(4-Fluorobenzyl)homopiperazine, 97%

Manufacturer: Alfa Aesar™ H5069406

Quantity 5g

 View more versions of this product

Catalog No. AAH5069406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76141-89-4
    Molecular Formula C12H17FN2
    Formula Weight 208.28
    MDL Number MFCD00816588
    UN Number UN2735
    Synonym 1-(4-Fluorobenzyl)-1,4-diazepane
    Chemical Name or Material 1-(4-Fluorobenzyl)homopiperazine

    Specifications

    Formula Weight 208.28
    Molecular Formula C12H17FN2
    MDL Number MFCD00816588
    CAS 76141-89-4
    UN Number UN2735
    Synonym 1-(4-Fluorobenzyl)-1,4-diazepane
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Quantity 5g
    Chemical Name or Material 1-(4-Fluorobenzyl)homopiperazine
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit