Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-fluorobenzyl)piperazine, 98%, Acros Organics

Click to view available options
Quantity:
1g
70931-28-1
C11H15FN2
194.253
MFCD01075234
OOSZCNKVJAVHJI-UHFFFAOYSA-N
1-4-fluorobenzyl piperazine,1-4-fluorophenyl methyl piperazine,1-4-fluorobenzyl-piperazine,piperazine,1-4-fluorophenyl methyl,1-4-fluoro-benzyl-piperazine,4-fluorobenzylpiperazine,piperazine, 1-4-fluorophenyl methyl,4-fluorophenyl methyl piperazine,4-fluorobenzyl-piperazine,acmc-1bl7k
796563
1-[(4-fluorophenyl)methyl]piperazine
C1CN(CCN1)CC2=CC=C(C=C2)F
70931-28-1
C11H15FN2
194.253
MFCD01075234
OOSZCNKVJAVHJI-UHFFFAOYSA-N
1-4-fluorobenzyl piperazine,1-4-fluorophenyl methyl piperazine,1-4-fluorobenzyl-piperazine,piperazine,1-4-fluorophenyl methyl,1-4-fluoro-benzyl-piperazine,4-fluorobenzylpiperazine,piperazine, 1-4-fluorophenyl methyl,4-fluorophenyl methyl piperazine,4-fluorobenzyl-piperazine,acmc-1bl7k
796563
1-[(4-fluorophenyl)methyl]piperazine
C1CN(CCN1)CC2=CC=C(C=C2)F
Quantity 1g
Refractive Index 1.5255 to 1.5275 (20°C, 589nm)
Color Yellow
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 194.25
Physical Form Liquid After Melting
Percent Purity ≥97.5%
Chemical Name or Material 1-(4-Fluorobenzyl)piperazine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger