Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluorophenyl)piperazine, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
2252-63-3
C10H13FN2
180.22
AVJKDKWRVSSJPK-UHFFFAOYSA-N
1-4-fluorophenyl piperazine,4-fluorophenylpiperazine,pfpp,4-fluoro-phenylpiperazine,n-4-fluorophenyl piperazine,para-fluorophenylpiperazine,unii-vml61be244,1-p-fluorophenyl piperazine,piperazine, 1-4-fluorophenyl,4-fluorophenyl piperazine
75260
1-(4-fluorophenyl)piperazine
C1CN(CCN1)C2=CC=C(C=C2)F
2252-63-3
C10H13FN2
180.22
AVJKDKWRVSSJPK-UHFFFAOYSA-N
1-4-fluorophenyl piperazine,4-fluorophenylpiperazine,pfpp,4-fluoro-phenylpiperazine,n-4-fluorophenyl piperazine,para-fluorophenylpiperazine,unii-vml61be244,1-p-fluorophenyl piperazine,piperazine, 1-4-fluorophenyl,4-fluorophenyl piperazine
75260
1-(4-fluorophenyl)piperazine
C1CN(CCN1)C2=CC=C(C=C2)F
Quantity 10g
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point >110°C
Color Yellow
Refractive Index 1.5585 to 1.5605
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 180.22
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material 1-(4-Fluorophenyl)piperazine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 218-846-6