Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Hydroxyphenyl)piperazine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
56621-48-8
C10H14N2O
178.235
MFCD00066156
GPEOAEVZTOQXLG-UHFFFAOYSA-N
1-4-hydroxyphenyl piperazine,4-piperazin-1-yl phenol,4-1-piperazinyl phenol,n-4-hydroxyphenyl piperazine,p-1-piperazinyl phenol,4-piperazinophenol,phenol, 4-1-piperazinyl,1-4-hydroxyphenyl-piperazine,4-piperazinylphenol,4-piperazinyl phenol
92467
4-piperazin-1-ylphenol
C1CN(CCN1)C2=CC=C(C=C2)O
56621-48-8
C10H14N2O
178.235
MFCD00066156
GPEOAEVZTOQXLG-UHFFFAOYSA-N
1-4-hydroxyphenyl piperazine,4-piperazin-1-yl phenol,4-1-piperazinyl phenol,n-4-hydroxyphenyl piperazine,p-1-piperazinyl phenol,4-piperazinophenol,phenol, 4-1-piperazinyl,1-4-hydroxyphenyl-piperazine,4-piperazinylphenol,4-piperazinyl phenol
92467
4-piperazin-1-ylphenol
C1CN(CCN1)C2=CC=C(C=C2)O
Packaging Glass Bottle
Infrared Spectrum Authentic
Melting Point 220.0°C
Color Brown
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 178.23
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 1-(4-Hydroxyphenyl)piperazine, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger