Alfa Aesar™ 1-(4-Iodophenyl)piperazine hydrochloride, 95%

Manufacturer: Alfa Aesar™ H5529903

Quantity 1g

 View more versions of this product

Catalog No. AAH5529903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 624726-35-8
    Formula Weight 324.59
    MDL Number MFCD04119724
    Chemical Name or Material 1-(4-Iodophenyl)piperazine hydrochloride

    Specifications

    Formula Weight 324.59
    Melting Point 177° to 181°C
    Physical Form Solid
    MDL Number MFCD04119724
    CAS 624726-35-8
    Quantity 1g
    Sensitivity Light Sensitive
    Chemical Name or Material 1-(4-Iodophenyl)piperazine hydrochloride
    Assay 95%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit