Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(4-Iodophenyl)pyrrole, 97%

Click to view available options
Quantity:
1g
5g
25g
92636-36-7
C10H8IN
269.085
MFCD00052399
FMURNAZHVQDQQN-UHFFFAOYSA-N
1-4-iodophenyl pyrrole,1-4-iodophenyl-1h-pyrrole,acmc-20apaj,4-iodophenyl pyrrole,maybridge1_002926,1-4-iodo-phenyl-1h-pyrrole,1h-pyrrole,1-4-iodophenyl,1-4-iodophenyl-1h-pyrrole #
272430
1-(4-iodophenyl)pyrrole
C1=CN(C=C1)C2=CC=C(C=C2)I
92636-36-7
C10H8IN
269.085
MFCD00052399
FMURNAZHVQDQQN-UHFFFAOYSA-N
1-4-iodophenyl pyrrole,1-4-iodophenyl-1h-pyrrole,acmc-20apaj,4-iodophenyl pyrrole,maybridge1_002926,1-4-iodo-phenyl-1h-pyrrole,1h-pyrrole,1-4-iodophenyl,1-4-iodophenyl-1h-pyrrole #
272430
1-(4-iodophenyl)pyrrole
C1=CN(C=C1)C2=CC=C(C=C2)I
Sensitivity Light sensitive
Quantity 1g
Melting Point 128°C to 133°C
Beilstein 7915717
Formula Weight 269.09
Percent Purity 97%
Chemical Name or Material 1-(4-Iodophenyl)pyrrole

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures