Alfa Aesar™ 1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene, 99+%

Manufacturer: Alfa Aesar™ H2748009

Quantity 10g

 View more versions of this product

Catalog No. AAH2748009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 39969-26-1
    Molecular Formula C18H18O
    Formula Weight 250.34
    MDL Number MFCD04038795
    Synonym 1-(4-Methoxyphenyl)-2-(4-n-propyl)phenylacetylene
    Chemical Name or Material 1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene

    Specifications

    Formula Weight 250.34
    Melting Point 61°C
    Molecular Formula C18H18O
    MDL Number MFCD04038795
    Boiling Point 162° to 164°C (1mmHg)
    CAS 39969-26-1
    Synonym 1-(4-Methoxyphenyl)-2-(4-n-propyl)phenylacetylene
    Quantity 10g
    Chemical Name or Material 1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit