Alfa Aesar™ 1-(4-Methylbenzyl)piperazine, 97%

Manufacturer: Alfa Aesar™ H5554803

Quantity 1g

 View more versions of this product

Catalog No. AAH5554803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 23173-57-1
    Molecular Formula C12H18N2
    Formula Weight 190.29
    MDL Number MFCD01075235
    Chemical Name or Material 1-(4-Methylbenzyl)piperazine

    Specifications

    Formula Weight 190.29
    MDL Number MFCD01075235
    Health Hazard 1 Danger
    Quantity 1g
    Flash Point 96°C (205°F)
    Health Hazard 2 GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C12H18N2
    CAS 23173-57-1
    Melting Point 37° to 41°C
    Chemical Name or Material 1-(4-Methylbenzyl)piperazine
    Assay 97%

    Description

    Applications
    is a useful synthetic intermediate in the synthesis of 3-Desmethyl 4-Methyl Meclizine Dihydrochloride ; a metabolite of Meclizine (, 2HCl monohydrate) which is an antiemetic. 1-(4-Methylbenzyl)piperazine is also used in the synthesis of inhibitors of acetylcholinesterase (AChE) and butyrylcholinesterase

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit