Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(4-Methylbenzyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5554806

Quantity 5g

 View more versions of this product

Catalog No. AAH5554806


  Check Availability
Add to cart

Specifications

Description

Applications
is a useful synthetic intermediate in the synthesis of 3-Desmethyl 4-Methyl Meclizine Dihydrochloride ; a metabolite of Meclizine (, 2HCl monohydrate) which is an antiemetic. 1-(4-Methylbenzyl)piperazine is also used in the synthesis of inhibitors of acetylcholinesterase (AChE) and butyrylcholinesterase

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

23173-57-1
190.29
1-(4-Methylbenzyl)piperazine
C12H18N2
MFCD01075235

Specifications

23173-57-1
190.29
37° to 41°C
96°C (205°F)
97%
C12H18N2
MFCD01075235
5g
1-(4-Methylbenzyl)piperazine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit