Alfa Aesar™ 1-(4-Methylphenyl)homopiperazine monohydrochloride, 98%

Manufacturer: Alfa Aesar™ H51750ME

Quantity 500mg

 View more versions of this product

Catalog No. AAH51750ME

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1192191-39-1
    Molecular Formula C12H18N2·HCl
    Formula Weight 226.75
    MDL Number MFCD16172201
    Beilstein 19720425
    Synonym 4-MPHP·HCl
    Chemical Name or Material 1-(4-Methylphenyl)homopiperazine monohydrochloride

    Specifications

    Formula Weight 226.75
    Molecular Formula C12H18N2·HCl
    MDL Number MFCD16172201
    CAS 1192191-39-1
    Beilstein 19720425
    Synonym 4-MPHP·HCl
    Melting Point 177° to 181°C
    Quantity 500mg
    Chemical Name or Material 1-(4-Methylphenyl)homopiperazine monohydrochloride
    Assay 98%

    Description

    Applications
    1-(4-Methylphenyl)homopiperazine monohydrochloride is useful intermediate for synthesis.

    Notes
    Sold in collaboration with Tosoh Organic Chemical Co., Ltd.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit