Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Nitrophenyl)piperazine 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
6269-89-2
C10H13N3O2
207.23
MFCD00005961
VWOJSRICSKDKAW-UHFFFAOYSA-N
1-4-nitrophenyl piperazine,1-4-nitro-phenyl-piperazine,piperazine, 1-4-nitrophenyl,1-4-nitrophenyl-piperazine,n-4-nitrophenyl piperazine,n-4-nitrophenyl-piperazine,4-nitrophenyl piperazine,1-4-nitrophenyl piperazin,pubchem8580,4-nitrophenylpiperazine
80447
C1CN(CCN1)C2=CC=C(C=C2)[N+](=O)[O-]
6269-89-2
C10H13N3O2
207.23
MFCD00005961
VWOJSRICSKDKAW-UHFFFAOYSA-N
1-4-nitrophenyl piperazine,1-4-nitro-phenyl-piperazine,piperazine, 1-4-nitrophenyl,1-4-nitrophenyl-piperazine,n-4-nitrophenyl piperazine,n-4-nitrophenyl-piperazine,4-nitrophenyl piperazine,1-4-nitrophenyl piperazin,pubchem8580,4-nitrophenylpiperazine
80447
C1CN(CCN1)C2=CC=C(C=C2)[N+](=O)[O-]
Color Gray to White
Quantity 25g
Melting Point 130.0°C to 134.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 207.23
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material 1-(4-Nitrophenyl)piperazine, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 228-443-7

TSCA : TSCA