Alfa Aesar™ 1,4-Oxathiane 4,4-dioxide, 99%

Manufacturer: Alfa Aesar™ A1021014

Quantity 25g

 View more versions of this product

Catalog No. AAA1021014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107-61-9
    Molecular Formula C4H8O3S
    Formula Weight 136.16
    MDL Number MFCD00011683
    Beilstein 110609
    Synonym 4,4-Dioxo-1,4-oxathiane; 1,4-Thioxane 1,1-dioxide
    Chemical Name or Material 1,4-Oxathiane 4,4-dioxide

    Specifications

    Formula Weight 136.16
    Synonym 4,4-Dioxo-1,4-oxathiane; 1,4-Thioxane 1,1-dioxide
    MDL Number MFCD00011683
    Beilstein 110609
    TSCA TSCA
    Molecular Formula C4H8O3S
    Odor Odorless
    RTECS Number RP4375000
    CAS 107-61-9
    Melting Point 131° to 133°C
    Quantity 25g
    Chemical Name or Material 1,4-Oxathiane 4,4-dioxide
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit