Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Oxathiane, 98+%

Click to view available options
Quantity:
5g
25g
15980-15-1
C4H8OS
104.167
MFCD00006573
JBYHSSAVUBIJMK-UHFFFAOYSA-N
1,4-thioxane,p-thioxane,1-oxa-4-thiacyclohexane,ccris 1746,1,4-oxathiin, 2,3,5,6-tetrahydro,thioather,1 pound not4-thioxane,1-thia-4-oxacyclohexane,acmc-1bt7b,wln: t6o dstj
27596
1,4-oxathiane
C1CSCCO1
15980-15-1
C4H8OS
104.167
MFCD00006573
JBYHSSAVUBIJMK-UHFFFAOYSA-N
1,4-thioxane,p-thioxane,1-oxa-4-thiacyclohexane,ccris 1746,1,4-oxathiin, 2,3,5,6-tetrahydro,thioather,1 pound not4-thioxane,1-thia-4-oxacyclohexane,acmc-1bt7b,wln: t6o dstj
27596
1,4-oxathiane
C1CSCCO1
Refractive Index 1.507
Density 1.114
Odor Unpleasant
Quantity 25g
Flash Point 42°C (107°F)
Boiling Point 147°C to 148°C
UN Number UN1993
Beilstein 102552
Formula Weight 104.17
Percent Purity ≥98%
Chemical Name or Material 1,4-Oxathiane
Show More Show Less

GHS H Statement
H226-H319
Flammable liquid and vapour.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 240-117-6

RTECSNumber : RP4200000

TSCA : Yes

Recommended Storage : Ambient temperatures