Learn More
eMolecules​ [1,4]Oxazepane | Oakwood Chemical | 5638-60-8 | MFCD09971804 | 101.149 | C5H11NO | 98.000 | C1CNCCOC1 | 5g | 537695590
Supplier:  eMolecules​ 537695590
Encompass_Preferred
[1,4]Oxazepane | Oakwood Chemical | 5638-60-8 | MFCD09971804 | 101.149 | C5H11NO | 98.000 | C1CNCCOC1 | 5g | 537695590
Catalog No. NC2285904