Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Pentadiene 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
591-93-5
C5H8
68.11
MFCD00008665
QYZLKGVUSQXAMU-UHFFFAOYSA-N
1,4-pentadiene,unii-a5if861oq5,pentadiene-1,4,allylethylene,penta-1,4-dien-1-yl,penta-1,4-dien-3-yl,ch2=chch2ch=ch2,acmc-1at58
11587
penta-1,4-diene
C=CCC=C
591-93-5
C5H8
68.11
MFCD00008665
QYZLKGVUSQXAMU-UHFFFAOYSA-N
1,4-pentadiene,unii-a5if861oq5,pentadiene-1,4,allylethylene,penta-1,4-dien-1-yl,penta-1,4-dien-3-yl,ch2=chch2ch=ch2,acmc-1at58
11587
penta-1,4-diene
C=CCC=C
Refractive Index 1.3884 to 1.3904
Specific Gravity 0.65
Boiling Point 26.0°C
Density 0.6500g/mL
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point −20°C
Melting Point -148.0°C
Color Colorless
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula H2C=CHCH2CH=CH2
Beilstein 01, 251
Fieser 01,783
Formula Weight 68.11
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1,4-Pentadiene, 99%
Show More Show Less

GHS P Statement
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-736-9

TSCA : TSCA