Alfa Aesar™ 1-(4-Pyridyl)pyridinium chloride hydrochloride, tech. 85%

Manufacturer: Alfa Aesar™ H5535106

Quantity 5g

 View more versions of this product

Catalog No. AAH5535106

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 5421-92-1
    Formula Weight 229.11
    MDL Number MFCD00012821
    Beilstein 3786515
    Chemical Name or Material 1-(4-Pyridyl)pyridinium chloride hydrochloride
    Grade Technical Grade

    Specifications

    Formula Weight 229.11
    Melting Point 166° to 170°C
    Sensitivity Hygroscopic
    Color Yellow to brown
    TSCA TSCA
    Physical Form Solid
    MDL Number MFCD00012821
    CAS 5421-92-1
    Beilstein 3786515
    Quantity 5g
    Chemical Name or Material 1-(4-Pyridyl)pyridinium chloride hydrochloride
    Assay 85%
    Grade Technical Grade

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit