Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[4-(Trifluoromethyl)-2-pyrimidinyl]-4-piperidinecarboxylic acid, ≥97%, Maybridge™

Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
465514-39-0
C11H12F3N3O2
275.231
MFCD02091615
QYAIISDWPUEPHG-UHFFFAOYSA-N
1-4-trifluoromethyl-2-pyrimidinyl-4-piperidinecarboxylic acid,1-4-trifluoromethyl pyrimidin-2-yl piperidine-4-carboxylic acid,1-4-trifluoromethyl-2-pyrimidinyl-4-piperidine carboxylic acid,1-4-trifluoromethyl pyrimid-2-yl piperidine-4-carboxylic acid,1-4-trifluoromethyl-pyrimidin-2-yl-4-piperidinecarboxylic acid
2777798
1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
C1CN(CCC1C(=O)O)C2=NC=CC(=N2)C(F)(F)F
465514-39-0
C11H12F3N3O2
275.231
MFCD02091615
QYAIISDWPUEPHG-UHFFFAOYSA-N
1-4-trifluoromethyl-2-pyrimidinyl-4-piperidinecarboxylic acid,1-4-trifluoromethyl pyrimidin-2-yl piperidine-4-carboxylic acid,1-4-trifluoromethyl-2-pyrimidinyl-4-piperidine carboxylic acid,1-4-trifluoromethyl pyrimid-2-yl piperidine-4-carboxylic acid,1-4-trifluoromethyl-pyrimidin-2-yl-4-piperidinecarboxylic acid
2777798
1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
C1CN(CCC1C(=O)O)C2=NC=CC(=N2)C(F)(F)F
Quantity 1g
Packaging Amber Glass Bottle
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 275.23
Percent Purity 97%
Chemical Name or Material 1-[4-(Trifluoromethyl)-2-pyrimidinyl]-4-piperidinecarboxylic acid

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.