Alfa Aesar™ 1-[4-(Trifluoromethyl)phenyl]propanol, 98%

Manufacturer: Alfa Aesar™ H3293603

Quantity 1g

 View more versions of this product

Catalog No. AAH3293603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67081-98-5
    Molecular Formula C10H11F3O
    Formula Weight 204.19
    MDL Number MFCD07773046
    Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]propanol

    Specifications

    Formula Weight 204.19
    MDL Number MFCD07773046
    Quantity 1g
    Molecular Formula C10H11F3O
    CAS 67081-98-5
    Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]propanol
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit