Alfa Aesar™ 1-[4-(Trifluoromethyl)phenyl]propanol, 98%

Manufacturer: Alfa Aesar™ H3293609

Quantity 10g

 View more versions of this product

Catalog No. AAH3293609

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 67081-98-5
    Molecular Formula C10H11F3O
    Formula Weight 204.19
    MDL Number MFCD07773046
    Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]propanol

    Specifications

    Formula Weight 204.19
    MDL Number MFCD07773046
    Quantity 10g
    Molecular Formula C10H11F3O
    CAS 67081-98-5
    Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]propanol
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit