Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5,5-Trimethylhydantoin 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
6851-81-6
C6H10N2O2
142.158
MFCD00040439
ZNYIPTYJBRGSSL-UHFFFAOYSA-N
1,5,5-trimethylhydantoin,3,4,4-trimethyl-2,5-dioxoimidazolidine,1,5,5-trimethyl-imidazolidine-2,4-dione,1,5,5-trimethyl-1,3-diazolidine-2,4-dione,hydantoin, 1,5,5-trimethyl,2,4-imidazolidinedione, 1,5,5-trimethyl,1,5,5 trimethylhydantoin,acmc-209o3u,1,5,5-trimethyl-hydantoin,1,5,5-trimethyl hydantoin
81295
1,5,5-trimethylimidazolidine-2,4-dione
CC1(C(=O)NC(=O)N1C)C
6851-81-6
C6H10N2O2
142.158
MFCD00040439
ZNYIPTYJBRGSSL-UHFFFAOYSA-N
1,5,5-trimethylhydantoin,3,4,4-trimethyl-2,5-dioxoimidazolidine,1,5,5-trimethyl-imidazolidine-2,4-dione,1,5,5-trimethyl-1,3-diazolidine-2,4-dione,hydantoin, 1,5,5-trimethyl,2,4-imidazolidinedione, 1,5,5-trimethyl,1,5,5 trimethylhydantoin,acmc-209o3u,1,5,5-trimethyl-hydantoin,1,5,5-trimethyl hydantoin
81295
1,5,5-trimethylimidazolidine-2,4-dione
CC1(C(=O)NC(=O)N1C)C
Melting Point 164°C
Quantity 25g
Color White
Formula Weight 142.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 1,5,5-Trimethylhydantoin