Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,5,9-Decatriene, cis + trans, 97%

Click to view available options
Quantity:
5mL
25mL
13393-64-1
C10H16
136.238
MFCD00055625
GMGYODAMPOELNZ-MDZDMXLPSA-N
1,5,9-decatriene,5e-deca-1,5,9-triene,1,5,9-decatriene, mixture of cis and trans,e-1,5,9-decatriene,5e-1,5,9-decatriene,5e-1,5,9-decatriene #,1,5,9-decatriene, cis + trans
5362636
(5E)-deca-1,5,9-triene
C=CCCC=CCCC=C
13393-64-1
C10H16
136.238
MFCD00055625
GMGYODAMPOELNZ-MDZDMXLPSA-N
1,5,9-decatriene,5e-deca-1,5,9-triene,1,5,9-decatriene, mixture of cis and trans,e-1,5,9-decatriene,5e-1,5,9-decatriene,5e-1,5,9-decatriene #,1,5,9-decatriene, cis + trans
5362636
(5E)-deca-1,5,9-triene
C=CCCC=CCCC=C
Density 0.765
Refractive Index 1.448
Boiling Point 54°C to 55°C (14mmHg)
Quantity 5mL
Flash Point 46°C (115°F)
UN Number UN3295
Formula Weight 136.24
Percent Purity 97%
Chemical Name or Material 1,5,9-Decatriene, cis + trans

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 236-474-2

TSCA : No

Recommended Storage : Ambient temperatures