Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,5,9-Triazacyclotridecane trihydrobromide, 98%

Click to view available options
Quantity:
100mg
500mg
74676-52-1
C10H26Br3N3
428.051
MFCD00013326
HRDCXXGINBVBMF-UHFFFAOYSA-N
1,5,9-triazacyclotridecane trihydrobromide,1,5,9-triazacyclotridecane-hydrogen bromide 1/3
24212082
1,5,9-triazacyclotridecane;trihydrobromide
C1CCNCCCNCCCNC1.Br.Br.Br
74676-52-1
C10H26Br3N3
428.051
MFCD00013326
HRDCXXGINBVBMF-UHFFFAOYSA-N
1,5,9-triazacyclotridecane trihydrobromide,1,5,9-triazacyclotridecane-hydrogen bromide 1/3
24212082
1,5,9-triazacyclotridecane;trihydrobromide
C1CCNCCCNCCCNC1.Br.Br.Br
Sensitivity Hygroscopic
Melting Point ~300°C (decomposition)
Quantity 500mg
Formula Weight 428.05
Percent Purity 98%
Chemical Name or Material 1,5,9-Triazacyclotridecane trihydrobromide

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures