Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(5-Chloro-2-hydroxy-4-methylphenyl)-3-phenyl-1,3-propanedione, 99%, Acros Organics

Click to view available options
Quantity:
1g
5067-23-2
C16H13ClO3
288.727
MFCD00209561
XNOUEBNIOPXNAO-UHFFFAOYSA-N
1-5-chloro-2-hydroxy-4-methylphenyl-3-phenylpropane-1,3-dione,1-5-chloro-2-hydroxy-4-methylphenyl-3-phenyl-1,3-propanedione,1,3-propanedione,1-5-chloro-2-hydroxy-4-methylphenyl-3-phenyl,1-2-hydroxy-4-methyl-5-chlorophenyl-3-phenyl-1,3-propanedione,1-5-chloro-2-hydroxy-4-methylphenyl-3-phenyl-1,3propanedione
736164
1-(5-chloro-2-hydroxy-4-methylphenyl)-3-phenylpropane-1,3-dione
CC1=C(C=C(C(=C1)O)C(=O)CC(=O)C2=CC=CC=C2)Cl
5067-23-2
C16H13ClO3
288.727
MFCD00209561
XNOUEBNIOPXNAO-UHFFFAOYSA-N
1-5-chloro-2-hydroxy-4-methylphenyl-3-phenylpropane-1,3-dione,1-5-chloro-2-hydroxy-4-methylphenyl-3-phenyl-1,3-propanedione,1,3-propanedione,1-5-chloro-2-hydroxy-4-methylphenyl-3-phenyl,1-2-hydroxy-4-methyl-5-chlorophenyl-3-phenyl-1,3-propanedione,1-5-chloro-2-hydroxy-4-methylphenyl-3-phenyl-1,3propanedione
736164
1-(5-chloro-2-hydroxy-4-methylphenyl)-3-phenylpropane-1,3-dione
CC1=C(C=C(C(=C1)O)C(=O)CC(=O)C2=CC=CC=C2)Cl
Infrared Spectrum Authentic
Quantity 1g
Color Yellow
Melting Point 123°C to 127°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5COCHCOC6H(Cl)(CH3)OH
Formula Weight 288.73
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 1-(5-Chloro-2-hydroxy-4-methylphenyl)-3-phenyl-1, 3-propanedione
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.