Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(5-Chloro-2-hydroxyphenyl)-3-phenyl-1,3-propanedione, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
5067-25-4
C15H11ClO3
274.7
MFCD00209562
VOVLKGWYQQCQHB-UHFFFAOYSA-N
1-5-chloro-2-hydroxyphenyl-3-phenylpropane-1,3-dione,1-5-chloro-2-hydroxyphenyl-3-phenyl-1,3-propanedione,1-5-chloranyl-2-oxidanyl-phenyl-3-phenyl-propane-1,3-dione,cambridge id 5479511,regid_for_cid_724088,1,3-propanedione,1-5-chloro-2-hydroxyphenyl-3-phenyl,1-2-hydroxy-5-chlorophenyl-3-phenyl-1,3-propanedione,1-5-chloro-2-hydroxy-phenyl-3-phenyl-propane-1,3-dione
724088
1-(5-chloro-2-hydroxyphenyl)-3-phenylpropane-1,3-dione
C1=CC=C(C=C1)C(=O)CC(=O)C2=C(C=CC(=C2)Cl)O
5067-25-4
C15H11ClO3
274.7
MFCD00209562
VOVLKGWYQQCQHB-UHFFFAOYSA-N
1-5-chloro-2-hydroxyphenyl-3-phenylpropane-1,3-dione,1-5-chloro-2-hydroxyphenyl-3-phenyl-1,3-propanedione,1-5-chloranyl-2-oxidanyl-phenyl-3-phenyl-propane-1,3-dione,cambridge id 5479511,regid_for_cid_724088,1,3-propanedione,1-5-chloro-2-hydroxyphenyl-3-phenyl,1-2-hydroxy-5-chlorophenyl-3-phenyl-1,3-propanedione,1-5-chloro-2-hydroxy-phenyl-3-phenyl-propane-1,3-dione
724088
1-(5-chloro-2-hydroxyphenyl)-3-phenylpropane-1,3-dione
C1=CC=C(C=C1)C(=O)CC(=O)C2=C(C=CC(=C2)Cl)O
Infrared Spectrum Authentic
Melting Point 107°C to 112°C
Color Yellow
Packaging Glass bottle
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula C6H5COCH2COC6H3(Cl)OH
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,ethanol,ethylacetate
Formula Weight 274.7
Physical Form Crystalline Needles
Percent Purity 98%
Chemical Name or Material 1-(5-Chloro-2-hydroxyphenyl)-3-phenyl-1, 3-propanedione
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning