Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Cyclooctadiene, 99+%, stabilized, purified by redistillation, AcroSeal™, Thermo Scientific™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
111-78-4
C8H12
108.18
MFCD00001752
VYXHVRARDIDEHS-QGTKBVGQSA-N
1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
10937607
(5Z)-cycloocta-1,5-diene
C1C\C=C/CC\C=C/1
111-78-4
C8H12
108.18
MFCD00001752
VYXHVRARDIDEHS-QGTKBVGQSA-N
1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
10937607
(5Z)-cycloocta-1,5-diene
C1C\C=C/CC\C=C/1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

CAS Min % 99.0
CAS Max % 100.0
Density 0.8800g/mL
Color Colorless
Melting Point -69.5°C
Boiling Point 151°C (101.3 kPa)
Flash Point 38°C
Assay Percent Range 99% min. (GC)
Infrared Spectrum Authentic
Beilstein 05,116
Packaging AcroSeal™ Glass Bottle
Merck Index 15,273
Refractive Index 1.4920 to 1.4940
Quantity 800mL
Solubility Information Solubility in water: 780mg/l (20°C). Other solubilities: soluble in benzene and carbon tetrachloride
Specific Gravity 0.88
Formula Weight 108.18
Physical Form Liquid
Percent Purity 99+%
Stabilizer 100 to 200ppm octadecyl 3-(3«, 5«-di-tert-butyl-4«-hydroxyphenyl)propionate
Viscosity 1.3 mPa.s (20°C)
Chemical Name or Material 1, 5-Cyclooctadiene
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Danger

EINECSNumber : 203-907-1

RUO – Research Use Only