Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) Hexafluorophosphate, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
38465-86-0
C34H38IrP2·F6P
845.81
MFCD00064800
MUHONFFTSOCMDH-JXNOXZOESA-N
1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate,1,5-cyclooctadiene bis methyldiphenylphosphine ir,1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate,1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate,iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate
5702663
(1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate
CP(C1=CC=CC=C1)C2=CC=CC=C2.CP(C1=CC=CC=C1)C2=CC=CC=C2.C1CC=CCCC=C1.F[P-](F)(F)(F)(F)F.[Ir]
38465-86-0
C34H38IrP2·F6P
845.81
MFCD00064800
MUHONFFTSOCMDH-JXNOXZOESA-N
1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate,1,5-cyclooctadiene bis methyldiphenylphosphine ir,1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate,1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate,iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate
5702663
(1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate
CP(C1=CC=CC=C1)C2=CC=CC=C2.CP(C1=CC=CC=C1)C2=CC=CC=C2.C1CC=CCCC=C1.F[P-](F)(F)(F)(F)F.[Ir]
Melting Point 224.0°C
Color Red
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min
Formula Weight 845.81
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material (1, 5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
Show More Show Less

GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do

GHS Signal Word: Warning