Alfa Aesar™ (1,5-Cyclooctadiene)dimethylplatinum(II), Pt 58.5%

Manufacturer: Alfa Aesar™ 13735MD

Quantity 250mg

 View more versions of this product

Catalog No. AA13735MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 12266-92-1
    Formula Weight 333.3
    MDL Number MFCD00064719
    Chemical Name or Material (1,5-Cyclooctadiene)dimethylplatinum(II)

    Specifications

    Formula Weight 333.3
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00064719
    Health Hazard 1 Danger
    TSCA TSCA
    Physical Form Powder
    CAS 12266-92-1
    Health Hazard 2 GHS H Statement
    H334-H335-H315-H319
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    Melting Point 103° to 105°C
    Concentration Pt 58.5%
    Quantity 250mg
    Chemical Name or Material (1,5-Cyclooctadiene)dimethylplatinum(II)

    GHS H Statement
    H334-H335-H315-H319
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit