Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Cyclooctadiene(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate, 99%

Catalog No. aa42057md
Click to view available options
Quantity:
250 mg

CAS: 64536-78-3 | C31H50F6IrNP2 | 804.90 g/mol

1,5-Cyclooctadiene(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate is used as a catalyst for hydrogenation of mono-, di-, tri-, and tetra-substituted substrates and the isomerization and hydroboration of alkenes. It is also used in isotope exchange reactions, especially the direct exchange of a hydrogen atom with its isotopes deuterium and tritium. Carbonyl groups are also known to direct the hydrogenation by the Crabtree catalyst is highly regioselective.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64536-78-3
Molecular Formula C31H50F6IrNP2
Molecular Weight (g/mol) 804.90
MDL Number MFCD00075097
InChI Key UJXHUUQZACSUOG-KJWGIZLLSA-N
Synonym crabtree's catalyst, unii-816rs2nbpn, 816rs2nbpn, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium hexafluorophosphate, iridium i hexafluorophosphate, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium i hexafluorophosphate, tricyclohexylphosphine 1,5-cyclooctadiene pyridine iridium i hexafluorophosphate, felkin-crabtree catalyst, crabtree's catalyst mi, crabtree's catalyst hexafluorophosphate
PubChem CID 5702647
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;iridium;pyridine;tricyclohexylphosphane;hexafluorophosphate
SMILES [Ir+].C1=CC=NC=C1.F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 175°C (decomposition)
Quantity 250 mg
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in acetone,dichloromethane,ethanol and diethyl ether; Insoluble in water.
Formula Weight 804.89
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Cyclooctadiene(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.