Alfa Aesar™ 1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate

Hydrogenation

Manufacturer: Alfa Aesar™ 1880902

Quantity 0.5g

 View more versions of this product

Catalog No. AA1880902

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38465-86-0
    Formula Weight 845.79
    MDL Number MFCD00064800
    Chemical Name or Material 1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate

    Specifications

    Formula Weight 845.79
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Sensitivity Air & Moisture Sensitive
    Physical Form Crystalline
    MDL Number MFCD00064800
    Solubility Information Soluble in acetone
    Health Hazard 1 Danger
    Melting Point 224°C (dec.)
    Quantity 0.5g
    CAS 38465-86-0
    Chemical Name or Material 1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit