Alfa Aesar™ 1,5-Diaminonaphthalene, 97%

Manufacturer: Alfa Aesar™ B2426206

Quantity 5g

 View more versions of this product

Catalog No. AAB2426206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2243-62-1
    Molecular Formula C10H10N2
    Formula Weight 158.2
    MDL Number MFCD00004029
    UN Number UN3077
    Beilstein 907947
    Synonym 1,5-Naphthalenediamine
    Chemical Name or Material 1,5-Diaminonaphthalene

    Specifications

    Formula Weight 158.2
    Molecular Formula C10H10N2
    MDL Number MFCD00004029
    UN Number UN3077
    Beilstein 907947
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H351
    Suspected of causing cancer.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Melting Point 189° to 192°C
    CAS 2243-62-1
    Synonym 1,5-Naphthalenediamine
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number QJ3400000
    Boiling Point 200° to 210°C (5mmHg)
    Quantity 5g
    Chemical Name or Material 1,5-Diaminonaphthalene
    Assay 97%

    Description

    Applications
    t is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. The reducing properties of 1,5-diaminonaphthalene (1,5-DAN) as a MALDI matrix ( matrix-assisted laser desorption ionization) to amino acid sequencing and disulfide bond mapping of human urotensin II possessing one disulfide bond, and human guanylin possessing two disulfide bonds.

    Solubility
    Soluble in water. (0.3 g/L)

    Notes
    Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H351
    Suspected of causing cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit