Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diaminopentane, 98%, Thermo Scientific Chemicals

Catalog Number AAB2303914
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
462-94-2
C5H14N2
102.18
MFCD00008239
VHRGRCVQAFMJIZ-UHFFFAOYSA-N
1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p
273
CHEBI:18127
pentane-1,5-diamine
NCCCCCN
This item is not returnable. View return policy
462-94-2
C5H14N2
102.18
MFCD00008239
VHRGRCVQAFMJIZ-UHFFFAOYSA-N
1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p
273
CHEBI:18127
pentane-1,5-diamine
NCCCCCN

1,5-Diaminopentane is mainly used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diaminopentane is mainly used in organic synthesis.

Solubility
Soluble in water.

Notes
Air sensitive & hygroscopic. Incompatible with acid chlorides, acids, acid anhydrides, strong oxidizing agents, carbon dioxide.
TRUSTED_SUSTAINABILITY
Melting Point 9°C
Density 0.873
Boiling Point 178°C to 180°C
Odor Amine-like
Assay Percent Range 0.98
Refractive Index 1.4585
Linear Formula H2N(CH2)5NH2
Quantity 25 g
UN Number UN2735
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,1609
Solubility Information Soluble in water.
Formula Weight 102.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,5-Diaminopentane
Show More Show Less
Hazard Category H227-H302+H312-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 207-329-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit