Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
5744-59-2
C6H8N2O2
140.142
MFCD00085040
PXRXGHUTKHXUGF-UHFFFAOYSA-N
1,5-dimethyl-1h-pyrazole-3-carboxylic acid,1,5-dimethyl-1h-pyrazole-3-carboxylicacid,1,5-dimethyl-3-pyrazolecarboxylic acid,1h-pyrazole-3-carboxylic acid, 1,5-dimethyl,1,5-dimethyl-1h-pyrazole-3-carboxylic,acmc-209lyb,pyrazole-3-carboxylic acid, 1,5-dimethyl,1h-pyrazole-3-carboxylicacid, 1,5-dimethyl,1,5-dimethyl-1h-pyrazole-3-carboxylic acid #,1,5-dimethyl-1h-pyrazole-3-carboxylic acid
587757
1,5-dimethylpyrazole-3-carboxylic acid
CC1=CC(=NN1C)C(=O)O
5744-59-2
C6H8N2O2
140.142
MFCD00085040
PXRXGHUTKHXUGF-UHFFFAOYSA-N
1,5-dimethyl-1h-pyrazole-3-carboxylic acid,1,5-dimethyl-1h-pyrazole-3-carboxylicacid,1,5-dimethyl-3-pyrazolecarboxylic acid,1h-pyrazole-3-carboxylic acid, 1,5-dimethyl,1,5-dimethyl-1h-pyrazole-3-carboxylic,acmc-209lyb,pyrazole-3-carboxylic acid, 1,5-dimethyl,1h-pyrazole-3-carboxylicacid, 1,5-dimethyl,1,5-dimethyl-1h-pyrazole-3-carboxylic acid #,1,5-dimethyl-1h-pyrazole-3-carboxylic acid
587757
1,5-dimethylpyrazole-3-carboxylic acid
CC1=CC(=NN1C)C(=O)O
Melting Point 170°C to 174°C
Quantity 25g
Beilstein 115917
Formula Weight 140.14
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 1,5-Dimethyl-1H-pyrazole-3-carboxylic acid

storageNote1 : 2° to 8°C