Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dimethyl-2-pyrrolecarbonitrile, 99%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass Bottle
56341-36-7
C7H8N2
120.155
MFCD00003086
DRXOPQFEWDRGKT-UHFFFAOYSA-N
1,5-dimethyl-1h-pyrrole-2-carbonitrile,1,5-dimethyl-2-pyrrolecarbonitrile,1h-pyrrole-2-carbonitrile, 1,5-dimethyl,2cyano-1,5-dimethylpyrrol,2-cyano-1,5-dimethylpyrrol,1,5-dimethyl-2-cyanopyrrole,1h-pyrrole-2-carbonitrile,1,5-dimethyl,1,5-dimethyl-1h-pyrrole-2-carbonitrile #,1h-pyrrole-2-carbonitrile, 1,5-dimethyl-9ci
91812
1,5-dimethylpyrrole-2-carbonitrile
CC1=CC=C(N1C)C#N
56341-36-7
C7H8N2
120.155
MFCD00003086
DRXOPQFEWDRGKT-UHFFFAOYSA-N
1,5-dimethyl-1h-pyrrole-2-carbonitrile,1,5-dimethyl-2-pyrrolecarbonitrile,1h-pyrrole-2-carbonitrile, 1,5-dimethyl,2cyano-1,5-dimethylpyrrol,2-cyano-1,5-dimethylpyrrol,1,5-dimethyl-2-cyanopyrrole,1h-pyrrole-2-carbonitrile,1,5-dimethyl,1,5-dimethyl-1h-pyrrole-2-carbonitrile #,1h-pyrrole-2-carbonitrile, 1,5-dimethyl-9ci
91812
1,5-dimethylpyrrole-2-carbonitrile
CC1=CC=C(N1C)C#N
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 50g
Color Brown-Gray
Melting Point 53°C to 56°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 120.15
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 1, 5-Dimethyl-2-pyrrolecarbonitrile, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 260-120-6

TSCA : TSCA