Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1,5-Diphenylcarbazide, ACS
Click to view available options
Quantity:
5g
25g
100g
Specifications
Sensitivity | Air and light sensitive |
Color | White to Cream |
Quantity | 5g |
Melting Point | 170°C to 176°C |
Beilstein | 752039 |
Merck Index | 14,3321 |
Solubility Information | Soluble in glacial acetic acid,alcohol and acetone; Slightly soluble in water. |
Formula Weight | 242.28 |
Physical Form | Crystalline |
Grade | ACS |
Show More |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-403-7
RTECSNumber : FF2750000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen