Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diphenylcarbohydrazide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
25g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

140-22-7
C13H14N4O
242.282
KSPIHGBHKVISFI-UHFFFAOYSA-N
8789
CHEBI:4641

High quality chemicals for laboratory and specialized industrial use

CAS 140-22-7
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C13H14N4O
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
IUPAC Name 1,3-dianilinourea
Molecular Weight (g/mol) 242.282
Quantity 25g
PubChem CID 8789
ChEBI CHEBI:4641
Formula Weight 242.28
Physical Form Powder
Chemical Name or Material 1,5-Diphenylcarbohydrazide
Grade Baker Analyzed™ ACS Reagent
Show More Show Less