Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,5-Naphthyridine, ≥97%

Click to view available options
Quantity:
250mg
1g
5g
254-79-5
C8H6N2
130.15
MFCD00039727
VMLKTERJLVWEJJ-UHFFFAOYSA-N
1,5-diazanaphthalene,1,5-pyridopyridine,pyridino 3,2-b pyridine,pyridopyridine,1,5-naphtyridine,1,5 naphthyridine,zlchem 698,pubchem18072,acmc-209gka,pyrido 1,5-b pyridine
136070
CHEBI:36625
1,5-naphthyridine
C1=CC2=C(C=CC=N2)N=C1
254-79-5
C8H6N2
130.15
MFCD00039727
VMLKTERJLVWEJJ-UHFFFAOYSA-N
1,5-diazanaphthalene,1,5-pyridopyridine,pyridino 3,2-b pyridine,pyridopyridine,1,5-naphtyridine,1,5 naphthyridine,zlchem 698,pubchem18072,acmc-209gka,pyrido 1,5-b pyridine
136070
CHEBI:36625
1,5-naphthyridine
C1=CC2=C(C=CC=N2)N=C1
Quantity 1g
Color White
Melting Point 69°C to 75°C
Density 1.25g/mL
Beilstein 109350
Formula Weight 130.15
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material 1,5-Naphthyridine

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger