Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(5-Nitrofurfurylidene)aminoguanidine Hydrochloride 99.0+%, TCI America™

Click to view available options
Quantity:
5g
946-48-5
C6H8ClN5O3
233.612
MFCD00142564
CHJIFAUFPHYNTD-JSGFVSQVSA-N
9570078
2-[(E)-(5-nitrofuran-2-yl)methylideneamino]guanidine;hydrochloride
C1=C(OC(=C1)[N+](=O)[O-])C=NN=C(N)N.Cl
946-48-5
C6H8ClN5O3
233.612
MFCD00142564
CHJIFAUFPHYNTD-JSGFVSQVSA-N
9570078
2-[(E)-(5-nitrofuran-2-yl)methylideneamino]guanidine;hydrochloride
C1=C(OC(=C1)[N+](=O)[O-])C=NN=C(N)N.Cl
Quantity 5g
Color Red-Yellow
Formula Weight 233.61
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (T)
Chemical Name or Material 1-(5-Nitrofurfurylidene)aminoguanidine Hydrochloride

RTECSNumber : MF4610000

TSCA : No