Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Pentanediol, 97%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Steel Pail
111-29-5
111-29-5
Packaging Steel Pail
Quantity 20 kg
CAS Max % 1
Percent Purity 0.97
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.