Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Pentanediol, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
5kg
Packaging:
Glass bottle
Metal drum
111-29-5
C5H12O2
104.15
ALQSHHUCVQOPAS-UHFFFAOYSA-N
1,5-pentanediol,1,5-dihydroxypentane,pentylene glycol,pentamethylene glycol,1,5-pentylene glycol,1,5 pentanediol,1,5-pentamethylene glycol,alpha,omega-pentanediol,unii-07uxz0scst,1,5-pentandiol
8105
pentane-1,5-diol
C(CCO)CCO
111-29-5
C5H12O2
104.15
ALQSHHUCVQOPAS-UHFFFAOYSA-N
1,5-pentanediol,1,5-dihydroxypentane,pentylene glycol,pentamethylene glycol,1,5-pentylene glycol,1,5 pentanediol,1,5-pentamethylene glycol,alpha,omega-pentanediol,unii-07uxz0scst,1,5-pentandiol
8105
pentane-1,5-diol
C(CCO)CCO
Viscosity 77 mPa.s (30°C)
Color Colorless
Quantity 500g
Flash Point 135°C
Melting Point -18.0°C
Density 0.9940g/mL
Packaging Glass bottle
Refractive Index 1.4490 to 1.4510
Specific Gravity 0.994
Boiling Point 242.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula HO(CH2)5OH
Beilstein 01,481
Merck Index 15,7229
Solubility Information Solubility in water: miscible. Other solubilities: limited solubility in benzene, trichloroethylene,, petroleum ether, heptane, miscible with alcohol, acetone and ethyl acetate, 11 g/110g ether (25°C)
Formula Weight 104.15
Physical Form Oily Liquid
Percent Purity 98%
Chemical Name or Material 1, 5-Pentanediol
Show More Show Less