Alfa Aesar™ 1,5-Pentanedithiol, 96%

Manufacturer: Alfa Aesar™ B2566506

Quantity 5g

 View more versions of this product

Catalog No. AAB2566506

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 928-98-3
    Molecular Formula HS(CH2)5SH
    Formula Weight 136.28
    MDL Number MFCD00004908
    UN Number UN3334
    Beilstein 1732335
    Synonym Pentamethylene dimercaptan
    Chemical Name or Material 1,5-Pentanedithiol

    Specifications

    Formula Weight 136.28
    MDL Number MFCD00004908
    UN Number UN3334
    Health Hazard 1 Warning
    Flash Point 95°C (203°F)
    Odor Unpleasant
    Melting Point −72°C
    Quantity 5g
    Molecular Formula HS(CH2)5SH
    CAS 928-98-3
    Beilstein 1732335
    Synonym Pentamethylene dimercaptan
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P280f-P403
    Wear protective clothing.
    Store in a well-ventilated place.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
    Boiling Point 216° to 218°C
    Chemical Name or Material 1,5-Pentanedithiol
    Assay 96%

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P280f-P403
    Wear protective clothing.
    Store in a well-ventilated place.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit