Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,5-Pentanedithiol, 96%

Manufacturer:  Alfa Aesar™ B2566514

Quantity 25g

 View more versions of this product

Catalog No. AAB2566514


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

928-98-3
136.28
UN3334
Pentamethylene dimercaptan
HS(CH2)5SH
MFCD00004908
1732335
1,5-Pentanedithiol

Specifications

95°C (203°F)
216° to 218°C
25g
HS(CH2)5SH
MFCD00004908
1732335
1,5-Pentanedithiol
Unpleasant
−72°C
928-98-3
136.28
UN3334
Pentamethylene dimercaptan
96%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P280f-P403
Wear protective clothing.
Store in a well-ventilated place.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit