Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate, 98+%, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
13242-55-2
C12H16O8
288.26
MFCD00063247
BAKQMOSGYGQJOJ-UHFZAUJKSA-N
triacetyllevoglucosan,1,6-anhydro-,a-d-glucopyranose 2,3,4-triacetate,2,3,4-tri-o-acetyl-1,6-anhydro-,a-d-glucopyranose,2r,3s,4r,5r-2,3-bis acetyloxy-6,8-dioxabicyclo 3.2.1 octan-4-yl acetate
134159065
[(1S,2S,3S,4S,5R)-3,4-diacetyloxy-6,8-dioxabicyclo[3.2.1]octan-2-yl] acetate
CC(=O)OC1C2COC(O2)C(C1OC(=O)C)OC(=O)C
13242-55-2
C12H16O8
288.26
MFCD00063247
BAKQMOSGYGQJOJ-UHFZAUJKSA-N
triacetyllevoglucosan,1,6-anhydro-,a-d-glucopyranose 2,3,4-triacetate,2,3,4-tri-o-acetyl-1,6-anhydro-,a-d-glucopyranose,2r,3s,4r,5r-2,3-bis acetyloxy-6,8-dioxabicyclo 3.2.1 octan-4-yl acetate
134159065
[(1S,2S,3S,4S,5R)-3,4-diacetyloxy-6,8-dioxabicyclo[3.2.1]octan-2-yl] acetate
CC(=O)OC1C2COC(O2)C(C1OC(=O)C)OC(=O)C
Infrared Spectrum Authentic
Color White
Specific Rotation − 66.00
Packaging Glass bottle
Melting Point 111.0°C to 112.0°C
Quantity 100mg
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Solubility Information Solubility in water: soluble.
Formula Weight 288.26
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate, 99%
Show More Show Less

EINECSNumber : 236-222-1

TSCA : TSCA