Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,6-Diacetoxyhexane, 98%

Click to view available options
Quantity:
50g
250g
6222-17-9
C10H18O4
202.25
MFCD00191655
ZMFWEWMHABZQNB-UHFFFAOYSA-N
1,6-diacetoxyhexane,1,6-hexanediol diacetate,hexylene glycol diacetate,1,6-hexanediol, diacetate,1,6-hexanediol, 1,6-diacetate,1,6-dihydroxyhexane diacetate,hexamethylene diacetate,hexane-1,6-diyl diacetate,acmc-209n0s,6-acetyloxy hexyl acetate
80356
6-acetyloxyhexyl acetate
CC(=O)OCCCCCCOC(=O)C
6222-17-9
C10H18O4
202.25
MFCD00191655
ZMFWEWMHABZQNB-UHFFFAOYSA-N
1,6-diacetoxyhexane,1,6-hexanediol diacetate,hexylene glycol diacetate,1,6-hexanediol, diacetate,1,6-hexanediol, 1,6-diacetate,1,6-dihydroxyhexane diacetate,hexamethylene diacetate,hexane-1,6-diyl diacetate,acmc-209n0s,6-acetyloxy hexyl acetate
80356
6-acetyloxyhexyl acetate
CC(=O)OCCCCCCOC(=O)C
Quantity 250g
Boiling Point 103°C to 105°C (6mmHg)
Formula Weight 202.25
Percent Purity 98%
Chemical Name or Material 1,6-Diacetoxyhexane