Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,6-Dibromoperfluorohexane, 96%

Click to view available options
Quantity:
1g
5g
918-22-9
C6Br2F12
459.855
MFCD00153110
SWAPKQWKFYIOJS-UHFFFAOYSA-N
1,6-dibromododecafluorohexane,1,6-dibromoperfluorohexane,hexane, 1,6-dibromo-dodecafluoro,dibromoperfluorohexane,acmc-209rdr,perfluoro 1,6-dibromohexane,hexane,1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro,1,6-bis bromanyl-1,1,2,2,3,3,4,4,5,5,6,6-dodecakis fluoranyl hexane
522992
1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexane
C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)Br)(F)F)(F)F
918-22-9
C6Br2F12
459.855
MFCD00153110
SWAPKQWKFYIOJS-UHFFFAOYSA-N
1,6-dibromododecafluorohexane,1,6-dibromoperfluorohexane,hexane, 1,6-dibromo-dodecafluoro,dibromoperfluorohexane,acmc-209rdr,perfluoro 1,6-dibromohexane,hexane,1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro,1,6-bis bromanyl-1,1,2,2,3,3,4,4,5,5,6,6-dodecakis fluoranyl hexane
522992
1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexane
C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)Br)(F)F)(F)F
Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Refractive Index 1.345
Quantity 5g
Boiling Point 141°C to 142°C
Beilstein 1822184
Solubility Information Soluble in water.
Formula Weight 459.87
Percent Purity 96%
Chemical Name or Material 1,6-Dibromoperfluorohexane

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures