Alfa Aesar™ 1,6-Dichlorohexane, 98%

Manufacturer: Alfa Aesar™ B2176322

Quantity 100g

 View more versions of this product

Catalog No. AAB2176322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2163-00-0
    Molecular Formula Cl(CH2)6Cl
    Formula Weight 155.07
    MDL Number MFCD00001020
    UN Number UN3082
    Beilstein 773673
    Synonym Hexamethylene dichloride
    Chemical Name or Material 1,6-Dichlorohexane

    Specifications

    Formula Weight 155.07
    TSCA TSCA
    Molecular Formula Cl(CH2)6Cl
    UN Number UN3082
    Beilstein 773673
    Flash Point 73°C (163°F)
    Melting Point −13°C
    Quantity 100g
    MDL Number MFCD00001020
    CAS 2163-00-0
    Synonym Hexamethylene dichloride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Boiling Point 204° to 205°C
    Chemical Name or Material 1,6-Dichlorohexane
    Assay 98%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit