Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,6-Dicyanohexane, 99%

Click to view available options
Quantity:
25g
100g
629-40-3
C8H12N2
136.198
MFCD00001981
BTNXBLUGMAMSSH-UHFFFAOYSA-N
1,6-dicyanohexane,suberonitrile,suberic acid dinitrile,suberodinitrile,unii-jnt1lxa02u,korksaeuredinitril german,jnt1lxa02u,hexamethylene di-isocyanide,hexane-1,6-dicarbonitrile,hexamethylenediisocyanic acid
12385
octanedinitrile
C(CCCC#N)CCC#N
629-40-3
C8H12N2
136.198
MFCD00001981
BTNXBLUGMAMSSH-UHFFFAOYSA-N
1,6-dicyanohexane,suberonitrile,suberic acid dinitrile,suberodinitrile,unii-jnt1lxa02u,korksaeuredinitril german,jnt1lxa02u,hexamethylene di-isocyanide,hexane-1,6-dicarbonitrile,hexamethylenediisocyanic acid
12385
octanedinitrile
C(CCCC#N)CCC#N
Quantity 25g
Flash Point 175°C (347°F)
Melting Point -3°C
Boiling Point 174°C to 176°C
UN Number UN3276
Beilstein 1744365
Formula Weight 136.2
Percent Purity 99%
Chemical Name or Material 1,6-Dicyanohexane

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

RTECSNumber : WM2113000

TSCA : TSCA