Alfa Aesar™ 1,6-Dicyanohexane, 99%

Manufacturer: Alfa Aesar™ B2454322

Quantity 100g

 View more versions of this product

Catalog No. AAB2454322

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 629-40-3
    Molecular Formula NC(CH2)6CN
    Formula Weight 136.2
    MDL Number MFCD00001981
    UN Number UN3276
    Beilstein 1744365
    Synonym Octanedinitrile; Suberonitrile
    Chemical Name or Material 1,6-Dicyanohexane

    Specifications

    Formula Weight 136.2
    Molecular Formula NC(CH2)6CN
    UN Number UN3276
    Beilstein 1744365
    Flash Point 175°C (347°F)
    Synonym Octanedinitrile; Suberonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    RTECS Number WM2113000
    Quantity 100g
    TSCA TSCA
    MDL Number MFCD00001981
    CAS 629-40-3
    Boiling Point 174° to 176°C
    Melting Point −3°C
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1,6-Dicyanohexane
    Assay 99%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit